Synonyms:
Methane dichlorideMethylene chloride
M0629-500ML
TCM0629-500MLEA
55.32
USD
InStock
TCM0629-500ML
Dichloromethane ≥99.5% stabilized
Dichloromethane
(stabilized with 2-Methyl-2-butene)
[for HPLC Solvent]
CAS Number: 75-09-2
MDL Number: MFCD00000881
Molecular Formula: CH2Cl2
Molecular Weight: 84.93
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 39
Melting point (°C): -97
Specific Gravity (20/20): 1.33
Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Methylene Chloride