Synonyms:
2-Methoxy-N-methylethanamine
M0986-25ML
M0986-5ML
TCM0986-25MLEA
310.59
USD
InStock
TCM0986-25ML
TCM0986-005ML
N-(2-Methoxyethyl)methylamine ≥97.0% (by GC, titration analysis)
N-(2-Methoxyethyl)methylamine
CAS Number: 38256-93-8
MDL Number: MFCD00144829
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 1
Specific Gravity (20/20): 0.83