Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Tolylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Synonyms: (4-Methylphenyl)metaboric acid 4-Methylbenzeneboronic acid 4-Methylphenylboronic acid Dihydroxy(4-methylphenyl)borane

M1126-5G M1126-25G
TCM1126-005GEA 75.09 USD InStock
TCM1126-005G TCM1126-25G
4-Tolylboronic acid (contains varying amounts of Anhydride)
4-Tolylboronic acid
CAS Number: 5720-05-8
MDL Number: MFCD00039138
Molecular Formula: C7H9BO2
Molecular Weight: 135.96
Form: Crystal
Color: White

Other Product Names / Synonyms: 4-Methylbenzeneboronic Acid, p-Tolylboronic Acid


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00039138
CAS Number: 5720-05-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/24/28974824.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty