Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mifepristone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: RU 486

M1732-1G M1732-5G
TCM1732-5GEA 732 USD InStock
TCM1732-5G TCM1732-1G
Mifepristone ≥98.0% (by HPLC, titration analysis)
Mifepristone
CAS Number: 84371-65-3
Molecular Formula: C29H35NO2
Molecular Weight: 429.60
Purity/Analysis Method: 98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 195
Specific rotation [a]20/D: 141 deg (C=0.5, CHCl3)
Storage Temperature: 0-10°C

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Steroids, Other Steroids
Other Product Names / Synonyms: 11beta-[4-(Dimethylamino)phenyl]-17beta-hydroxy-17-(1-propynyl)estra-4,9-dien-3-one


Formula: C₂₉H₃₅NO₂
MW: 429.60 g/mol
Melting Pt: 195 °C
CAS Number: 84371-65-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/60/28976060.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T,HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty