M1956-200MG
M1956-1G
TCM1956-200MGEA
81.17
USD
InStock
TCM1956-200MG
TCM1956-1G
Mono-2-O-(p-toluenesulfonyl)-β-cyclodextrin ≥98.0% (by HPLC)
Mono-2-O-(p-toluenesulfonyl)-alpha-cyclodextrin
CAS Number: 93184-10-2
Molecular Formula: C43H66O32S
Molecular Weight: 1127.03
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 176
Specific rotation [a]20/D: 116 deg (C=4, DMSO)