Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrothiophenol ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 1-Mercapto-4-nitrobenzene4-Nitrobenzene-1-thiolp-Nitrothiophenol4-Nitrobenzenethiol

N0146-5G N0146-25G
TCN0146-5GEA 69.59 USD InStock
TCN0146-5G TCN0146-025G
4-Nitrothiophenol ≥95.0% (by titrimetric analysis)
4-Nitrothiophenol
CAS Number: 1849-36-1
MDL Number: MFCD00007343
Molecular Formula: C6H5NO2S
Molecular Weight: 155.17
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 79

Other Product Names / Synonyms: 4-Nitrothiophenol


Formula: C₆H₅NO₂S
Melting Pt: 79 °C
MDL Number: MFCD00007343
CAS Number: 1849-36-1
EINECS: 217-436-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/42/28989242.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty