Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutral red

Supplier: TCI America
Synonyms: N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride Basic Red 5

N0315-25G
TCN0315-025GEA 75.13 USD InStock
TCN0315-025G
Neutral red
Neutral red
CAS Number: 553-24-2
MDL Number: MFCD00012651
Molecular Formula: C15H17ClN4
Molecular Weight: 288.78
Form: Crystal
Color: Deep Green
Melting point (°C): 290

Other Product Names / Synonyms: Neutral Red, Toluylene Red


Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/92/28989392.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty