N0392-10MG
TCN0392-010MGEA
36.54
USD
InStock
TCN0392-010MG
4-Nitrophenyl alpha-L-Fucopyranoside ≥98.0% (by HPLC)
4-Nitrophenyl-alpha-L-fucopyranoside
CAS Number: 10231-84-2
MDL Number: MFCD00063697
Molecular Formula: C12H15NO7
Molecular Weight: 285.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C