Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophenyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

N0619-1G
TCN0619-001GEA 176.61 USD InStock
TCN0619-001G
4-Nitrophenyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)
4-Nitrophenyl-alpha-D-mannopyranoside
[Substrate for alpha-Mannosidase]
CAS Number: 10357-27-4
MDL Number: MFCD00066002
Molecular Formula: C12H15NO8
Molecular Weight: 301.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 180
Specific rotation [a]20/D: 160 deg (C=0.2, H2O)


Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00066002
CAS Number: 10357-27-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/17/28989817.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty