Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Azacytidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-oneLadakamycin

A2033-1G A2033-100MG
TCA2033-1GEA 205.03 USD InStock
TCA2033-1G TCA2033-100MG
5-Azacytidine ≥98.0% (by titrimetric analysis)
5-Azacytidine
CAS Number: 320-67-2
MDL Number: MFCD00006539
Molecular Formula: C8H12N4O5
Molecular Weight: 244.21
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 216
Storage Temperature: 0°C

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Epigenetics Resea
Other Product Names / Synonyms: 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, Ladakamycin


Formula: C₈H₁₂N₄O₅
MW: 244.21 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00006539
CAS Number: 320-67-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/01/80/28930180.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty