Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorokerosene, super-high boiling

Supplier: TCI America
Synonyms: PFK

P1034-5G
TCP1034-005GEA 152.19 USD InStock
TCP1034-005G
Perfluorokerosene, super-high boiling
Perfluorokerosene
[for Mass spectrometry]
MDL Number: MFCD00132835
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 2.05

Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials


https://us-legacy.vwr.com/stibo/web/std.lang.all/16/68/28991668.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty