Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorotriamylamine (so called)

Supplier: TCI America
Synonyms: Perfluorotripentylamine

P1051-25G
TCP1051-25GEA 65.9 USD InStock
TCP1051-25G
Perfluorotriamylamine (so called)
Perfluorotriamylamine
CAS Number: 338-84-1
MDL Number: MFCD00042367
Molecular Formula: C15F33N
Molecular Weight: 821.12
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.94

Other Product Names / Synonyms: Perfluorotripentylamine


Formula: C₁₅F₃₃N
MW: 821.12 g/mol
Density: 1.94 (20 °C)
MDL Number: MFCD00042367
CAS Number: 338-84-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/16/86/28991686.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty