Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorokerosene 50% in PFK low boiling, super-high boiling

Supplier: TCI America

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: PFK

P1061-5G
TCP1061-005GEA 285.09 USD InStock
TCP1061-005G
Perfluorokerosene 50% in PFK low boiling, super-high boiling
Perfluorokerosene
[for Mass spectrometry]
MDL Number: MFCD00132835
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 46

Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials


https://us-legacy.vwr.com/stibo/web/std.lang.all/16/96/28991696.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty