Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Phenoxyvaleric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1357-5G
TCP1357-005GEA 545.83 USD InStock
TCP1357-005G
5-Phenoxyvaleric acid ≥98.0% (by titrimetric analysis)
5-Phenoxyvaleric acid
CAS Number: 7170-40-3
MDL Number: MFCD02093487
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 165
Melting point (°C): 63


Formula: C₁₁H₁₄O₃
Boiling Pt: 165 °C
Melting Pt: 63 °C
MDL Number: MFCD02093487
CAS Number: 7170-40-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/05/28992005.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty