Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose ≥97.0%

Supplier: TCI America
Synonyms: (3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetatealpha,beta-D-Mannose pentaacetate

P1514-5G
TCP1514-5GEA 77.14 USD InStock
TCP1514-5G
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose ≥97.0%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
CAS Number: 25941-03-1
MDL Number: MFCD05864874
Molecular Formula: C16H22O11
Molecular Weight: 390.34
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 68
Specific rotation [a]20/D: 52 deg (C=1, CHCl3)
Storage Temperature: 0-10°C


Formula: C₁₆H₂₂O₁₁
MW: 390.34 g/mol
Melting Pt: 68 °C
MDL Number: MFCD05864874
CAS Number: 25941-03-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/45/28992145.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty