Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperacillin sodium salt ≥98.0% (by total nitrogen basis)

Supplier: TCI America

P1774-1G P1774-5G
TCP1774-5GEA 331.81 USD InStock
TCP1774-5G TCP1774-1G
Piperacillin sodium salt ≥98.0% (by total nitrogen basis)
Piperacillin sodium salt
CAS Number: 59703-84-3
MDL Number: MFCD00056867
Molecular Formula: C23H27N5O7S
Molecular Weight: 539.54
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 185
Specific rotation [a]20/D: 180 deg (C=0.8, H2O)


Formula: C₂₃H₂₇N₅O₇S
Melting Pt: 185 °C
MDL Number: MFCD00056867
CAS Number: 59703-84-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/80/28993080.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty