Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium 0.1 mmol/g on SH silica gel

Supplier: TCI America

P1786-1G
TCP1786-1GEA 389 USD InStock
TCP1786-1G
Palladium 0.1 mmol/g on SH silica gel
Palladium
CAS Number:
Molecular Formula:
Molecular Weight:
Form: Crystals
Color: Yellow

Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis


Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty