Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ribonucleic acid (from yeast)

Supplier: TCI America
Synonyms: Nucleic Acid

R0022-25G
TCR0022-025GEA 85.97 USD InStock
TCR0022-025G
Ribonucleic acid (from yeast)
Ribonucleic acid
MDL Number: MFCD00132195
Form: Crystal
Color: White

Other Product Names / Synonyms: Nucleic Acid


MDL Number: MFCD00132195
CAS Number: 63231-63-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty