Would you like to log in again?
DTT / TG12 [Dithiothreitol + α-thioglycerol] (1:2 mixture)
Supplier: TCI America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Synonyms: Dithiothreitol + alpha-Thioglycerol
S0437-1G
TCS0437-001GEA 123 USD InStock
TCS0437-001G
DTT / TG12 [Dithiothreitol + α-thioglycerol] (1:2 mixture)
DTT / TG12 [Dithiothreitol + alpha-thioglycerol] (1:2 mixture)
[Matrix for FABMS and liquid SIMS]
MDL Number: MFCD01321186
Other Product Names / Synonyms: Dithiothreitol + alpha-Thioglycerol
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty