Synonyms:
Terpinolene
T0817-25ML
T0817-500ML
TCT0817-025MLEA
27.19
USD
InStock
TCT0817-025ML
TCT0817-500ML
Terpinolene (p-mentha-1,4(8)-diene) ≥85.0%
Terpinolene (p-Mentha-1,4(8)-diene)
CAS Number: 586-62-9
MDL Number: MFCD00049191
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: 85.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 185
Flash Point (°C): 37
Specific Gravity (20/20): 0.86