Synonyms:
Tetraethylgermanium
T1157-1G
TCT1157-001GEA
29.42
USD
InStock
TCT1157-001G
Tetraethylgermane ≥98.0%
Tetraethylgermane
CAS Number: 597-63-7
MDL Number: MFCD00015096
Molecular Formula: C8H20Ge
Molecular Weight: 188.88
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 35
Specific Gravity (20/20): 1.00