Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylphosphonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) complex ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1416-1G
TCT1416-001GEA 384.58 USD InStock
TCT1416-001G
Tetrabutylphosphonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) complex ≥98.0% (by titrimetric analysis)
Tetrabutylphosphonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) complex
CAS Number: 105029-70-7
MDL Number: MFCD00191494
Molecular Formula: C22H36NiPS10
Molecular Weight: 710.80
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Deep Green


Formula: C₂₂H₃₆NiPS₁₀
MDL Number: MFCD00191494
CAS Number: 105029-70-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/38/28995438.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty