Synonyms:
(S)-(-)-2-Amino-3-(3-indolyl)propanol
T1537-1G
TCT1537-001GEA
92.67
USD
InStock
TCT1537-001G
L-(-)-Tryptophanol ≥97.0% (by titrimetric analysis)
L-(-)-Tryptophanol
CAS Number: 2899-29-8
MDL Number: MFCD00037970
Molecular Formula: C11H14N2O
Molecular Weight: 190.25
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -21 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Amino-3-(3-indolyl)propanol