Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Triisopropylsilyl)indole ≥94.0%

Supplier: TCI America

T1822-1G
TCT1822-001GEA 302 USD InStock
TCT1822-001G
1-(Triisopropylsilyl)indole ≥94.0%
1-(Triisopropylsilyl)indole
CAS Number: 123191-00-4
MDL Number: MFCD02093498
Molecular Formula: C17H27NSi
Molecular Weight: 273.50
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Color: Yellow
Specific Gravity (20/20): 0.99


Formula: C₁₇H₂₇NSi
Density: 0.99 (20 °C)
MDL Number: MFCD02093498
CAS Number: 123191-00-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/98/28995798.jpg
Order Now
Specification Test Results
Purity/Analysis Method 94.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty