Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(8-quinolinolato)aluminum ≥98.0% (by titrimetric analysis), purified by sublimation

Supplier: TCI America
Synonyms: 8-Quinolinol Aluminum ComplexAlq3Aluminum triquinolin-8-olateAluminum Tris(8-hydroxyquinolinate)

T2238-5G
TCT2238-5GEA 257 USD InStock
TCT2238-5G
Tris(8-quinolinolato)aluminum ≥98.0% (by titrimetric analysis), purified by sublimation
Tris(8-quinolinolato)aluminum
CAS Number: 2085-33-8
MDL Number: MFCD00191693
Molecular Formula: C27H18AlN3O3
Molecular Weight: 459.44
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 418

Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Highly Purified Reagents
Other Product Names / Synonyms: Aluminum Tris(8-hydroxyquinolinate), 8-Quinolinol Aluminum Complex, Alq3


Formula: C₂₇H₁₈AlN₃O₃
MW: 459.44 g/mol
Melting Pt: 418 °C
MDL Number: MFCD00191693
CAS Number: 2085-33-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/45/28996045.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty