Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mannose triflate ≥98.0%

Supplier: TCI America
Synonyms: TATM(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose

T2307-100MG
TCT2307-100MGEA 49 USD InStock
TCT2307-100MG
Mannose triflate ≥98.0%
Mannose triflate
CAS Number: 92051-23-5
Molecular Formula: C15H19F3O12S
Molecular Weight: 480.36
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 122
Specific rotation [a]20/D: -16 deg (C=1, CH3CN)
Storage Temperature: 0°C

Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Bioscience, Mannose, O-Substituted Sugars
Other Product Names / Synonyms: TATM


Formula: C₁₅H₁₉F₃O₁₂S
MW: 480.36 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00012353
CAS Number: 92051-23-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/88/28996088.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty