T2384-25G
T2384-5G
TCT2384-25GEA
219
USD
InStock
TCT2384-25G
TCT2384-5G
2-(Trifluoromethyl)thioxanthen-9-one ≥98.0%
2-(Trifluoromethyl)thioxanthen-9-one
CAS Number: 1693-28-3
MDL Number: MFCD00079785
Molecular Formula: C14H7F3OS
Molecular Weight: 280.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 149