Synonyms:
(3,4-Dimethoxyphenyl)methanol
, Veratryl alcohol
V0020-25ML
TCV0020-025MLEA
50.75
USD
InStock
TCV0020-025ML
3,4-Dimethoxybenzyl alcohol ≥98.0%
3,4-Dimethoxybenzyl alcohol
CAS Number: 93-03-8
MDL Number: MFCD00004638
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 173
Flash Point (°C): 113
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 3,4-Dimethoxybenzyl Alcohol