Synonyms:
NBD-F
A5593-100MG
TCA5593-100MGEA
291.07
USD
InStock
TCA5593-100MG
4-Fluoro-7-nitrobenzofurazan ≥99.0% (by HPLC)
4-Fluoro-7-nitrobenzofurazan
[for HPLC Labeling]
CAS Number: 29270-56-2
MDL Number: MFCD00010196
Molecular Formula: C6H2FN3O3
Molecular Weight: 183.10
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 110
Storage Temperature: 0°C
Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Fluoro-7-nitrobenzofurazan, 4-Fluoro-7-nitro-2,1,3-benzoxadiazole