Would you like to log in again?
Stirring Shaft, Cresent Type, Ace Glass Incorporated
Supplier: Ace Glass
8073-16 8073-23 8073-19
89061-956EA 139.55 USD InStock
89061-956 89061-954 89061-958
Stirring Shaft, Cresent Type, Ace Glass Incorporated
Stirrers Stirring Shafts
Precision ground glass stirring shafts, 10mm O.D., crescent type
Useful for stirring viscous materials at low rpm. Available for round bottom flasks of 1, 3 and 5 liters. Catelog #89061-956, three liter size, will also fit a two liter flask, but closely. Flask opening must be a [ST] 29/42 joint or larger. Use with Trubore® bearings. Compatible with Ace PTFE mechanical seal and PTFE collet-type bearings. Join to stirrer motor using a chuck. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty