Would you like to log in again?
E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Supplier: Omega Bio-Tek
E.Z.N.A®
D3392-01 D3392-00 D3392-00 D3392-01 D3392-02 D3392-02
101319-412EA 116.06 USD InStock
CA101319-436 101319-412 CA101319-348 101319-436 101319-348 CA101319-412
E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Blood DNA Kit provides rapid total DNA isolation from 1–250 µL of fresh and frozen anticoagulated whole blood
This kit allows for simultaneous processing of single or multiple samples in less than 30 minutes. Phenol/chloroform extractions, and time-consuming steps such as precipitation with isopropanol or ethanol have been eliminated. DNA purified with the E.Z.N.A.® Blood DNA method is ready for applications such as PCR, Southern blotting, or restriction enzyme digestion.
The Kit can also be used for the preparation of genomic DNA from buffy coat, serum, plasma, bone marrow, lymphocytes, platelets, and body fluids.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty