Would you like to log in again?
E-Z 96® Plant DNA Kit, Omega Bio-tek®
Supplier: Omega Bio-Tek
E-Z 96®
D1086-01 D1086-02 D1086-01 D1086-02
101318-936EA 277.53 USD InStock
101318-936 CA101318-938 CA101318-936 101318-938
E-Z 96® Plant DNA Kit, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E-Z 96® Plant DNA Kit has adapted the buffer system from Omega Bio-tek®'s SP Plant DNA system
This is particularly useful to process varieties of plants, such as those with unusually high levels of phenolic compounds or polysaccharides as exemplified in cotton, pine, and peanut samples. Up to 50mg of wet tissue or 12mg of dry tissue can be processed in each well in less than 1 hour. The system combines the reversible nucleic acid-binding properties of the HiBind® matrix with the speed and versatility of the E-Z 96® DNA plate to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. Purified DNA is suitable for PCR, restriction digestion, and hybridization techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty