Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-fluoroaniline ≥97%

Supplier: Matrix Scientific

002083-25G 002083-100G
101775-746EA 58.46 USD InStock
101775-746 101775-744
3-Chloro-2-fluoroaniline ≥97%
3-Chloro-2-fluoroaniline
TCA listed: T
Hazard: Toxic


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 214 °C
MDL Number: MFCD00069415
CAS Number: 2106-04-9
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 214°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty