Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-o-toluidine ≥99%

Supplier: Matrix Scientific
Synonyms: 5-Fluoro-2-methylaniline2-Amino-4-fluorotoluene

002155-100G
101775-810EA 60 USD InStock
101775-810
5-Fluoro-o-toluidine ≥99%
5-Fluoro-o-toluidine
Hazard: Toxic/Irritant/Store in Freezer


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 98…100 °C (15 mmHg)
Melting Pt: 38…40 °C
MDL Number: MFCD00007764
CAS Number: 367-29-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 98-100°/15
ND20 1.538

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty