Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-fluoroaniline ≥97%

Supplier: Matrix Scientific

002689-25G 002689-100G
101776-288EA 138.46 USD InStock
101776-288 101776-286
2-Chloro-5-fluoroaniline ≥97%
2-Chloro-5-fluoroaniline
Hazard: Toxic


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Melting Pt: ∼ 26 °C
MDL Number: MFCD00084852
CAS Number: 452-83-5
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Melting point ca 26°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty