Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylselenocyanate

Supplier: Matrix Scientific

003185-1G
101776-522EA 30.77 USD InStock
101776-522
Phenylselenocyanate
Phenylselenocyanate
Hazard: Toxic


Formula: C₇H₅NSe
Boiling Pt: 116…117 °C (12 mmHg)
MDL Number: MFCD00216944
CAS Number: 2179-79-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty