Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylvaleric acid ≥99%

Supplier: Matrix Scientific
Synonyms: Isocaproic acid4-Methylpentanoic acid

003196-100G 003196-25G 003196-5G
101776-530EA 73.85 USD InStock
101776-530 101776-532 101776-528
4-Methylvaleric acid ≥99%
4-Methylvaleric acid
TCA listed: T
Hazard: Corrosive/Stench


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 199…201 °C
Melting Pt: –35 °C
Density: 0.923
MDL Number: MFCD00002803
CAS Number: 646-07-1
Order Now
Specification Test Results
Boiling point 199-201°
Density 0.923
ND20 1.4146

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty