Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzylamine ≥97%

Supplier: Matrix Scientific
Synonyms: (4-Fluorophenyl)methanamine

003348-500G 003348-100G 003348-25G
101776-676EA 330.77 USD InStock
101776-676 101776-672 101776-674
4-Fluorobenzylamine ≥97%
4-Fluorobenzylamine
TCA listed: T
Hazard: Corrosive/Air Sensitive


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 183 °C
Density: 1.095
MDL Number: MFCD00008120
CAS Number: 140-75-0
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Boiling point 183°
Density 1.095
ND20 1.512

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty