Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzylamine ≥97%

Supplier: Matrix Scientific

003349-100G 003349-25G
101776-678EA 100 USD InStock
101776-678 101776-680
2-Fluorobenzylamine ≥97%
2-Fluorobenzylamine
Hazard: Corrosive/Air Sensitive


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 73…75 °C (13 mmHg)
Density: 1.095
MDL Number: MFCD00008107
CAS Number: 89-99-6
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Boiling point 73-75°/13m
Density 1.095
ND20 1.517

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty