Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene-α-sulfonyl chloride ≥98%

Supplier: Matrix Scientific
Synonyms: α-Toluenesulfonyl chlorideα-Toluenesulphonyl chloridePhenylmethylsulfonyl chloridePhenylmethylsulphonyl chloridePhenylmethanesulfonyl chlorideBenzylsulphonyl chloridePhenylmethanesulphonyl chloride

003833-100G
101777-168EA 260 USD InStock
101777-168
Toluene-α-sulfonyl chloride ≥98%
Toluene-α-sulphonyl chloride
TCA listed: T
Hazard: Corrosive/Moisture Sensitive


Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD00007455
CAS Number: 1939-99-7
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Melting point 92-94°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty