Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Fluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific
Synonyms: 2-Fluorobenzyl cyanide2-(2-Fluorophenyl)acetonitrile

003901-25G 003901-100G
101777-222EA 21.54 USD InStock
101777-222 101777-224
(2-Fluorophenyl)acetonitrile ≥97%
(2-Fluorophenyl)acetonitrile
TCA listed: T
Hazard: Toxic/Irritant


Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 128…132 °C (20 mmHg)
Density: 1.059
MDL Number: MFCD00001897
CAS Number: 326-62-5
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 128-132°/2
Density 1.059
ND20 1.5009

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty