Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Diethylamino)sulfur trifluoride ≥97%

Supplier: Matrix Scientific
Synonyms: DAST

004186-25G 004186-100G 004186-5G
101777-420EA 838.46 USD InStock
101777-420 101777-418 101777-416
(Diethylamino)sulfur trifluoride ≥97%
(Diethylamino)sulphur trifluoride
Store in HDPE bottles in Freezer!

Hazard: Flammable/Corrosive/Store in Freezer


Formula: C₄H₁₀F₃NS
MW: 161.19 g/mol
Boiling Pt: 30…32 °C (3 mmHg)
Density: 1.22
Flash Pt: 23 °C
MDL Number: MFCD00000363
CAS Number: 38078-09-0
UN: 2920
ADR: 8,I
Order Now
Specification Test Results
Boiling point 30-32°/3mm
Density 1.22

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty