Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoaniline ≥98%

Supplier: Matrix Scientific

004327-25G 004327-100G
101777-496EA 100 USD InStock
101777-496 101777-494
2-Iodoaniline ≥98%
2-Iodoaniline
TCA listed: T
Hazard: Toxic/Irritant/Light Sensitive


Formula: C₆H₆IN
MW: 219.02 g/mol
Melting Pt: 55…58 °C
MDL Number: MFCD00007680
CAS Number: 615-43-0
Order Now
Specification Test Results
Melting point 55-58°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty