Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercaptobenzyl alcohol ≥98%

Supplier: Matrix Scientific

004838-25G
101777-786EA 807.69 USD InStock
101777-786
2-Mercaptobenzyl alcohol ≥98%
2-Mercaptobenzyl alcohol
Hazard: Toxic/Stench/Refrigerate


Formula: C₇H₈OS
MW: 140.21 g/mol
Melting Pt: 31…32 °C
MDL Number: MFCD00014448
CAS Number: 4521-31-7
Order Now
Specification Test Results
Melting point 31-32°
ND20 1.6231

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty