Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific
Synonyms: 2,4-Difluorobenzene-1-sulfonyl chloride2,4-Difluorobenzene-1-sulphonyl chloride

005182-100G
101778-060EA 113.85 USD InStock
101778-060
2,4-Difluorobenzenesulfonyl chloride ≥97%
2,4-Difluorobenzenesulphonyl chloride
Hazard: Corrosive/Irritant


Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 101…102 °C (1 mmHg)
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 101-102°/1

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty