Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Difluorobenzene ≥98%

Supplier: Matrix Scientific

005188-500G
101778-080EA 136.92 USD InStock
101778-080
1,2-Difluorobenzene ≥98%
1,2-Difluorobenzene
Hazard: Flammable


Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 92 °C
Melting Pt: –34 °C
Density: 1.158
MDL Number: MFCD00000284
CAS Number: 367-11-3
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Boiling point 92°
Density 1.158
ND20 1.443

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty