Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Difluoro-4-nitrobenzene ≥98%

Supplier: Matrix Scientific
Synonyms: 3,4-Difluoronitrobenzene

005547-500G
101778-474EA 169.23 USD InStock
101778-474
1,2-Difluoro-4-nitrobenzene ≥98%
1,2-Difluoro-4-nitrobenzene
Hazard: Flammable


Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 76…80 °C (11 mmHg)
Density: 1.437
MDL Number: MFCD00007198
CAS Number: 369-34-6
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 76-80°/11m
Density 1.437
ND20 1.509

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty