Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α,α',α',α'-Hexafluoro-p-xylene ≥98%

Supplier: Matrix Scientific
Synonyms: 1,4-Bis(trifluoromethyl)benzeneα,α,α,β,β,β-Hexafluoro-p-xylene

005867-500G 005867-100G
101778-698EA 43.08 USD InStock
101778-698 101778-700
α,α,α,α',α',α'-Hexafluoro-p-xylene ≥98%
α,α,α,α',α',α'-Hexafluoro-p-xylene
TCA listed: T
Hazard: Flammable


Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 116 °C
Density: 1.381
MDL Number: MFCD00000402
CAS Number: 433-19-2
Order Now
Specification Test Results
Boiling point 116°
Density 1.381
ND20 1.379

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty