Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Difluorobiphenyl ≥98%

Supplier: Matrix Scientific
Synonyms: PFB No. 154,4-Difluoro-1,1-biphenyl

006471-5G 006471-100G
101779-118EA 211.11 USD InStock
101779-118 101779-114
4,4'-Difluorobiphenyl ≥98%
4,4'-Difluorobiphenyl
Hazard: Irritant-Harmful


Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 254…255 °C
Melting Pt: 88…91 °C
MDL Number: MFCD00000349
CAS Number: 398-23-2
Order Now
Specification Test Results
Boiling point 254-255°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty