Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dimethylimidazole-4-sulfonyl chloride ≥95%

Supplier: Matrix Scientific
Synonyms: 1,2-Dimethyl-1H-imidazole-4-sulphonyl chloride

007142-1G
101779-404EA 60 USD InStock
101779-404
1,2-Dimethylimidazole-4-sulfonyl chloride ≥95%
1,2-Dimethylimidazole-4-sulphonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₅H₇ClN₂O₂S
Melting Pt: 91…94 °C
MDL Number: MFCD01313780
CAS Number: 137049-02-6
Order Now
Specification Test Results
Melting point 91-94°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty